Assay Details
Gene Symbol: LOC5577130 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021856919.1 | XP_021712611.1 | 7 - 8 | 2323 | 95 | |
XM_021856923.1 | XP_021712615.1 | 6 - 7 | 2419 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
