Assay Details
Gene Symbol: LOC5571700 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659794.2 | XP_001659844.2 | 2 - 3 | 1682 | 97 | |
XM_021842003.1 | XP_021697695.1 | 2 - 3 | 1690 | 97 |
