Assay Details
Gene Symbol: LOC5572351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021841121.1 | XP_021696813.1 | 7 - 8 | 2464 | 61 | |
XM_021841122.1 | XP_021696814.1 | 8 - 9 | 2094 | 61 | ||
XM_021841123.1 | XP_021696815.1 | 8 - 9 | 1985 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
