Assay Details
Gene Symbol: LOC5568352 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838248.1 | XP_021693940.1 | 8 | 6745 | 70 | |
XM_021838249.1 | XP_021693941.1 | 8 | 6742 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
