Assay Details
Gene Symbol: LOC5564062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648318.2 | XP_001648368.1 | 2 | 454 | 75 | |
GenBank mRNA | AY433069.1 | - | 2 | 426 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
