Assay Details
Gene Symbol: LOC5580010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653840.2 | XP_001653890.2 | 2 | 439 | 108 | |
GenBank mRNA | AY433190.1 | - | 2 | 318 | 108 | |
AY626233.1 | - | 1 | 310 | 108 | ||
CR938245.1 | - | 2 | 318 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
