Assay Details
Gene Symbol: LOC110680777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021856561.1 | XP_021712253.1 | 3 | 872 | 106 | |
GenBank mRNA | CR937623.1 | - | 2 | 618 | 106 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
