Assay Details
Gene Symbol: LOC5564556 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648846.2 | XP_001648896.1 | 4 | 1073 | 59 | |
GenBank mRNA | AY431452.1 | - | 2 | 419 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
