Assay Details
Gene Symbol: LOC5564080 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021857264.1 | XP_021712956.1 | 1 | 259 | 101 | |
XM_021857265.1 | XP_021712957.1 | 1 | 259 | 101 | ||
XM_021857266.1 | XP_021712958.1 | 1 | 259 | 101 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021857264.1, XM_021857265.1, XM_021857266.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
