Assay Details
Gene Symbol: LOC5576559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850178.1 | XP_021705870.1 | 12 - 13 | 5646 | 90 | |
XM_021850186.1 | XP_021705878.1 | 12 - 13 | 4242 | 90 | ||
XM_021850189.1 | XP_021705881.1 | 11 - 12 | 5595 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
