Assay Details
Gene Symbol: LOC5576559 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021850178.1 | XP_021705870.1 | 4 - 5 | 2257 | 77 | |
XM_021850186.1 | XP_021705878.1 | 4 - 5 | 853 | 77 | ||
XM_021850189.1 | XP_021705881.1 | 4 - 5 | 2257 | 77 | ||
XM_021850196.1 | XP_021705888.1 | 4 - 5 | 2257 | 77 | ||
XM_021850206.1 | XP_021705898.1 | 4 - 5 | 2257 | 77 | ||
XM_021850211.1 | XP_021705903.1 | 4 - 5 | 2257 | 77 | ||
XM_021850218.1 | XP_021705910.1 | 4 - 5 | 2257 | 77 | ||
XM_021850224.1 | XP_021705916.1 | 4 - 5 | 2257 | 77 |
