Assay Details
Gene Symbol: LOC5579655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648079.2 | XP_001648129.1 | 4 - 5 | 485 | 71 | |
XM_021852485.1 | XP_021708177.1 | 4 - 5 | 387 | 71 | ||
XM_021852486.1 | XP_021708178.1 | 4 - 5 | 314 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
