Assay Details
Gene Symbol: LOC5579655 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648079.2 | XP_001648129.1 | 7 - 8 | 1142 | 66 | |
XM_021852485.1 | XP_021708177.1 | 7 - 8 | 1044 | 66 | ||
XM_021852486.1 | XP_021708178.1 | 7 - 8 | 971 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
