Assay Details
Gene Symbol: LOC5568531 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852096.1 | XP_021707788.1 | 7 - 8 | 3983 | 118 | |
XM_021852100.1 | XP_021707792.1 | 7 - 8 | 3983 | 118 |

More Information
Set Membership: |
![]() ![]() ![]() |
