Assay Details
Gene Symbol: LOC5578258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663732.2 | XP_001663782.1 | 1 - 2 | 356 | 64 | |
XM_021841793.1 | XP_021697485.1 | 2 - 3 | 213 | 64 |

More Information
Set Membership: |
![]() ![]() |
