Assay Details
Gene Symbol: LOC5578258 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663732.2 | XP_001663782.1 | 2 - 3 | 1214 | 85 | |
XM_021841793.1 | XP_021697485.1 | 3 - 4 | 1071 | 85 | ||
XM_021841794.1 | XP_021697486.1 | 2 - 3 | 1022 | 85 |
