Assay Details
Gene Symbol: LOC5576358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837605.1 | XP_021693297.1 | 3 - 4 | 1108 | 80 | |
XM_021837606.1 | XP_021693298.1 | 3 - 4 | 1108 | 80 |
