Assay Details
Gene Symbol: LOC5576358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021837605.1 | XP_021693297.1 | 4 - 5 | 1387 | 67 | |
XM_021837606.1 | XP_021693298.1 | 4 - 5 | 1387 | 67 |

More Information
Set Membership: |
![]() ![]() |
