Assay Details
Gene Symbol: LOC5577665 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021838126.1 | XP_021693818.1 | 2 - 3 | 1396 | 61 | |
XM_021838128.1 | XP_021693820.1 | 2 - 3 | 1396 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
