Assay Details
Gene Symbol: LOC5568590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652372.2 | XP_001652422.2 | 8 - 9 | 2256 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5568590 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652372.2 | XP_001652422.2 | 8 - 9 | 2256 | 59 |
Set Membership: |
![]() ![]() ![]() ![]() |