Assay Details
Gene Symbol: LOC5576415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656067.2 | XP_001656117.2 | 1 - 2 | 829 | 73 | |
XM_001656068.2 | XP_001656118.2 | 1 - 2 | 230 | 73 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001656067.2, XM_001656068.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
