Assay Details
Gene Symbol: LOC5576415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656067.2 | XP_001656117.2 | 2 - 3 | 941 | 90 | |
XM_001656068.2 | XP_001656118.2 | 2 - 3 | 342 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
