Assay Details
Gene Symbol: LOC5576415 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656067.2 | XP_001656117.2 | 3 - 4 | 1218 | 85 | |
XM_001656068.2 | XP_001656118.2 | 3 - 4 | 619 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
