Assay Details
Gene Symbol: LOC5576440 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001656082.2 | XP_001656132.1 | 3 - 4 | 500 | 65 | |
XM_001656084.2 | XP_001656134.1 | 3 - 4 | 487 | 65 | ||
XM_011495371.2 | XP_011493673.1 | 3 - 4 | 447 | 65 | ||
XM_021846050.1 | XP_021701742.1 | 3 - 4 | 452 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
