Assay Details
Gene Symbol: LOC5566122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839662.1 | XP_021695354.1 | 11 - 12 | 2154 | 115 | |
XM_021839663.1 | XP_021695355.1 | 10 - 11 | 2013 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() |
