Assay Details
Gene Symbol: LOC5566122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839662.1 | XP_021695354.1 | 2 - 3 | 423 | 100 | |
XM_021839665.1 | XP_021695357.1 | 2 - 3 | 423 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
