Assay Details
Gene Symbol: LOC5566122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839662.1 | XP_021695354.1 | 8 - 9 | 1691 | 69 | |
XM_021839663.1 | XP_021695355.1 | 7 - 8 | 1550 | 69 | ||
XM_021839665.1 | XP_021695357.1 | 8 - 9 | 1691 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
