Assay Details
Gene Symbol: LOC409390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ698737.1 | - | 94 | |||
HP492021.1 | - | 94 | ||||
HP565229.1 | - | 94 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC409390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AJ698737.1 | - | 94 | |||
HP492021.1 | - | 94 | ||||
HP565229.1 | - | 94 |