Assay Details
Gene Symbol: Ac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077808.1 | NP_001071276.1 | 67 | |||
GenBank mRNA | AJ968562.1 | - | 67 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ac3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077808.1 | NP_001071276.1 | 67 | |||
GenBank mRNA | AJ968562.1 | - | 67 |