Assay Details
Gene Symbol: Amih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011568.1 | NP_001011568.1 | 90 | |||
GenBank mRNA | AY280848.1 | - | 90 | |||
AY739658.1 | - | 90 | ||||
AY739659.1 | - | 90 |
