Assay Details
Gene Symbol: Sod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178027.1 | NP_001171498.1 | 100 | |||
GenBank mRNA | AY329355.1 | - | 100 | |||
BI515107.1 | - | 100 | ||||
DB743032.1 | - | 100 | ||||
HP462474.1 | - | 100 | ||||
HP503564.1 | - | 100 |
