Assay Details
Gene Symbol: Sod2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178048.2 | NP_001171519.1 | 122 | |||
GenBank mRNA | AY329356.1 | - | 122 | |||
HP469582.1 | - | 122 | ||||
HP545478.1 | - | 122 |
