Assay Details
Gene Symbol: MsrA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178047.1 | NP_001171518.1 | 112 | |||
GenBank mRNA | AY329360.1 | - | 112 | |||
HP479842.1 | - | 112 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MsrA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001178047.1 | NP_001171518.1 | 112 | |||
GenBank mRNA | AY329360.1 | - | 112 | |||
HP479842.1 | - | 112 |