Assay Details
Gene Symbol: TyHyd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011633.1 | NP_001011633.1 | 105 | |||
GenBank mRNA | AY855337.1 | - | 105 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: TyHyd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011633.1 | NP_001011633.1 | 105 | |||
GenBank mRNA | AY855337.1 | - | 105 |