Assay Details
Gene Symbol: LOC551621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098219.1 | NP_001091689.1 | 80 | |||
GenBank mRNA | HP565328.1 | - | 80 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC551621 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098219.1 | NP_001091689.1 | 80 | |||
GenBank mRNA | HP565328.1 | - | 80 |