Assay Details
Gene Symbol: Hbg3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001011608.1 | NP_001011608.1 | 131 | |||
GenBank mRNA | D79208.1 | - | 131 | |||
HP499841.1 | - | 131 | ||||
HP557041.1 | - | 131 |
