Assay Details
Gene Symbol: Stip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080205.1 | NP_001073674.1 | 108 | |||
GenBank mRNA | DQ342041.1 | - | 108 | |||
HP504433.1 | - | 108 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Stip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080205.1 | NP_001073674.1 | 108 | |||
GenBank mRNA | DQ342041.1 | - | 108 | |||
HP504433.1 | - | 108 |