Assay Details
Gene Symbol: Mad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042458.1 | NP_001035923.1 | 91 | |||
GenBank mRNA | DQ666693.1 | - | 91 | |||
HP488817.1 | - | 91 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Mad | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042458.1 | NP_001035923.1 | 91 | |||
GenBank mRNA | DQ666693.1 | - | 91 | |||
HP488817.1 | - | 91 |