Assay Details
Gene Symbol: Grd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305884.1 | NP_001292813.1 | 87 | |||
GenBank mRNA | DQ667183.1 | - | 87 | |||
KJ485711.1 | - | 87 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Grd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305884.1 | NP_001292813.1 | 87 | |||
GenBank mRNA | DQ667183.1 | - | 87 | |||
KJ485711.1 | - | 87 |