Assay Details
Gene Symbol: Grd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001305884.1 | NP_001292813.1 | 102 | |||
GenBank mRNA | DQ667183.1 | - | 102 | |||
HP502593.1 | - | 102 | ||||
KJ485711.1 | - | 102 |
