Assay Details
Gene Symbol: pHCl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001143878.1 | NP_001137350.1 | 62 | |||
NM_001143879.1 | NP_001137351.1 | 62 | ||||
NM_001143880.1 | NP_001137352.1 | 62 | ||||
GenBank mRNA | DQ667189.1 | - | 62 | |||
DQ667191.1 | - | 62 | ||||
DQ667192.1 | - | 62 |
