Assay Details
Gene Symbol: Amel_8916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077822.1 | NP_001071290.1 | 109 | |||
GenBank mRNA | DQ667193.1 | - | 109 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Amel_8916 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077822.1 | NP_001071290.1 | 109 | |||
GenBank mRNA | DQ667193.1 | - | 109 |