Assay Details
Gene Symbol: Dcr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123013.2 | NP_001116485.2 | 86 | |||
GenBank mRNA | EU520658.1 | - | 86 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dcr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001123013.2 | NP_001116485.2 | 86 | |||
GenBank mRNA | EU520658.1 | - | 86 |