Assay Details
Gene Symbol: LOC411586 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP488952.1 | - | 99 | |||
HP488953.1 | - | 99 | ||||
HP488954.1 | - | 99 | ||||
HP488955.1 | - | 99 | ||||
HP488956.1 | - | 99 | ||||
HP488957.1 | - | 99 | ||||
HP510415.1 | - | 99 | ||||
HP510416.1 | - | 99 | ||||
HP510417.1 | - | 99 | ||||
HP510418.1 | - | 99 | ||||
HP510419.1 | - | 99 | ||||
HP510420.1 | - | 99 | ||||
HP510421.1 | - | 99 | ||||
HP510422.1 | - | 99 | ||||
HP510423.1 | - | 99 | ||||
HP510424.1 | - | 99 | ||||
HP510425.1 | - | 99 | ||||
HP510426.1 | - | 99 | ||||
HP510427.1 | - | 99 | ||||
HP510428.1 | - | 99 | ||||
HP510429.1 | - | 99 | ||||
HP510430.1 | - | 99 | ||||
HP540556.1 | - | 101 | ||||
HP551746.1 | - | 99 | ||||
HP565905.1 | - | 99 | ||||
HP565906.1 | - | 99 | ||||
HP565907.1 | - | 99 |
