Assay Details
Gene Symbol: LOC726262 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001327964.1 | NP_001314893.1 | 73 | |||
GenBank mRNA | HE611296.1 | - | 73 | |||
HP485623.1 | - | 73 | ||||
HP485624.1 | - | 73 |
