Assay Details
Gene Symbol: LOC413891 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP466871.1 | - | 75 | |||
HP476354.1 | - | 75 | ||||
HP476355.1 | - | 75 | ||||
HP501166.1 | - | 75 | ||||
HP518468.1 | - | 75 | ||||
HP537524.1 | - | 75 | ||||
HP544893.1 | - | 75 | ||||
HP544894.1 | - | 75 | ||||
HP568179.1 | - | 75 |
