Assay Details
Gene Symbol: LOC413005 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP477260.1 | - | 116 | |||
HP511498.1 | - | 116 | ||||
HP511499.1 | - | 116 | ||||
HP511500.1 | - | 116 | ||||
HP511501.1 | - | 116 | ||||
HP511502.1 | - | 116 | ||||
HP511503.1 | - | 116 | ||||
HP511504.1 | - | 116 | ||||
HP511505.1 | - | 116 | ||||
HP511506.1 | - | 116 | ||||
HP511507.1 | - | 116 | ||||
HP511508.1 | - | 116 | ||||
HP511509.1 | - | 116 | ||||
HP511510.1 | - | 116 | ||||
HP511511.1 | - | 116 | ||||
HP511512.1 | - | 116 | ||||
HP511513.1 | - | 116 | ||||
HP511514.1 | - | 116 | ||||
HP511515.1 | - | 116 | ||||
HP528664.1 | - | 116 | ||||
HP528666.1 | - | 116 | ||||
HP528668.1 | - | 116 | ||||
HP528670.1 | - | 116 | ||||
HP528672.1 | - | 116 | ||||
HP528675.1 | - | 116 | ||||
HP528677.1 | - | 116 | ||||
HP528679.1 | - | 116 | ||||
HP528681.1 | - | 116 | ||||
HP528683.1 | - | 116 | ||||
HP528685.1 | - | 116 | ||||
HP528687.1 | - | 116 | ||||
HP544683.1 | - | 116 | ||||
HP544684.1 | - | 116 | ||||
HP544685.1 | - | 116 | ||||
HP544686.1 | - | 116 | ||||
HP544687.1 | - | 116 | ||||
HP544688.1 | - | 116 | ||||
HP559779.1 | - | 116 | ||||
HP559780.1 | - | 116 |
