Assay Details
Gene Symbol: LOC413005 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP477260.1 | - | 76 | |||
HP511498.1 | - | 76 | ||||
HP511499.1 | - | 76 | ||||
HP511500.1 | - | 76 | ||||
HP511507.1 | - | 76 | ||||
HP511508.1 | - | 76 | ||||
HP511509.1 | - | 76 | ||||
HP528679.1 | - | 76 | ||||
HP528680.1 | - | 76 | ||||
HP528681.1 | - | 76 | ||||
HP528682.1 | - | 76 | ||||
HP528683.1 | - | 76 | ||||
HP528684.1 | - | 76 | ||||
HP528686.1 | - | 76 | ||||
HP528687.1 | - | 76 | ||||
HP544685.1 | - | 76 | ||||
HP544687.1 | - | 76 | ||||
HP544688.1 | - | 76 | ||||
HP559780.1 | - | 76 |
