Assay Details
Gene Symbol: LOC727552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP493815.1 | - | 95 | |||
HP493816.1 | - | 95 | ||||
HP574289.1 | - | 95 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC727552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | HP493815.1 | - | 95 | |||
HP493816.1 | - | 95 | ||||
HP574289.1 | - | 95 |