Assay Details
Gene Symbol: Rps14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001242469.1 | NP_001229398.1 | 123 | |||
GenBank mRNA | HP476951.1 | - | 123 | |||
HP499332.1 | - | 123 | ||||
HP532474.1 | - | 123 |
